siRNA Details
Gene Symbol: Galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024373.1 | NP_077349.1 | 2 | 279 | ||
XM_006254471.2 | XP_006254533.1 | 3 | 1700 | |||
GenBank mRNA | U35890.1 | - | 2 | 279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
