siRNA Details
Gene Symbol: Aco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024398.2 | NP_077374.2 | 2 | 105 | ||
GenBank mRNA | AJ243266.1 | - | 2 | 106 | ||
BC061999.1 | - | 2 | 105 | 5621226 | ||
FQ216755.1 | - | 2 | 130 |
