siRNA Details
Gene Symbol: Erap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030836.1 | NP_110463.1 | 1 | 358 | ||
XM_006231700.1 | XP_006231762.1 | 2 | 1092 | |||
XM_006231701.2 | XP_006231763.1 | 2 | 411 | |||
XM_006231704.2 | XP_006231766.1 | 2 | 411 | |||
XM_008760600.1 | XP_008758822.1 | 1 | 2504 | |||
XM_008760601.1 | XP_008758823.1 | 1 | 2505 | |||
XM_008760602.1 | XP_008758824.1 | 1 | 2503 | |||
GenBank mRNA | AF148323.1 | - | 1 | 355 | ||
AF148324.1 | - | 1 | 358 | |||
BC098664.1 | - | 2 | 365 | 7192921 |
