siRNA Details
Gene Symbol: Avil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024401.2 | NP_077377.2 | 4 | 316 | ||
XM_008765423.1 | XP_008763645.1 | 3 | 448 | |||
GenBank mRNA | AF099929.1 | - | 4 | 305 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Avil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024401.2 | NP_077377.2 | 4 | 316 | ||
XM_008765423.1 | XP_008763645.1 | 3 | 448 | |||
GenBank mRNA | AF099929.1 | - | 4 | 305 |