siRNA Details
Gene Symbol: PHKG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000294.2 | NP_000285.1 | 10 | 1486 | ||
GenBank mRNA | AB209055.1 | - | 5 | 875 | ||
AK289492.1 | - | 10 | 1350 | |||
BC002541.2 | - | 10 | 1346 | 3138927 | ||
M31606.1 | - | 10 | 1369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
