siRNA Details
Gene Symbol: Erap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030836.1 | NP_110463.1 | 6 | 1164 | ||
XM_006231700.1 | XP_006231762.1 | 7 | 1898 | |||
XM_006231701.2 | XP_006231763.1 | 7 | 1217 | |||
XM_006231704.2 | XP_006231766.1 | 7 | 1217 | |||
XM_008760600.1 | XP_008758822.1 | 6 | 3310 | |||
XM_008760601.1 | XP_008758823.1 | 6 | 3311 | |||
XM_008760602.1 | XP_008758824.1 | 6 | 3309 | |||
GenBank mRNA | AF148323.1 | - | 6 | 1161 | ||
AF148324.1 | - | 6 | 1164 | |||
BC098664.1 | - | 7 | 1171 | 7192921 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030711, AK159406, AK163954, AB047552, AK029841, and others.
Gene Ontology Categories:
