siRNA Details
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024268.2 | NP_001019439.1 | 3 | 240 | ||
GenBank mRNA | AF276774.1 | - | 3 | 255 | ||
BC093604.1 | - | 3 | 224 | 7317599 | ||
FQ132357.1 | - | 3 | 240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
