siRNA Details
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270575.1 | NP_001257504.1 | 4 | 351 | ||
NM_001270576.1 | NP_001257505.1 | 4 | 263 | |||
NM_030991.3 | NP_112253.1 | 4 | 351 | |||
GenBank mRNA | AB003991.1 | - | 142 | |||
AB003992.1 | - | 3 | 142 | |||
AF245227.1 | - | 4 | 350 | |||
BC087699.1 | - | 4 | 306 | 7314318 | ||
DY315301.1 | - | 4 | 283 | |||
FM033263.1 | - | 4 | 263 | |||
FQ212040.1 | - | 4 | 322 | |||
FQ213289.1 | - | 4 | 319 | |||
FQ213923.1 | - | 4 | 322 | |||
FQ214162.1 | - | 4 | 351 | |||
U56261.1 | - | 115 | ||||
U56262.1 | - | 3 | 115 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011428, M22012, AF483516, BC018249, AK078038, and others.
Gene Ontology Categories:
