siRNA Details
Gene Symbol: Adcy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031007.1 | NP_112269.1 | 2, 3 | 482 | ||
GenBank mRNA | M80550.1 | - | 2, 3 | 482 |

More Information
Other Species Targeted

Gene Ontology Categories:
