siRNA Details
Gene Symbol: Anpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031012.1 | NP_112274.1 | 1 | 132 | ||
XM_006229407.2 | XP_006229469.1 | 2 | 552 | |||
XM_006229408.2 | XP_006229470.1 | 2 | 573 | |||
XM_006229409.1 | XP_006229471.1 | 2 | 609 | |||
GenBank mRNA | AF039891.1 | - | 2 | 614 | ||
M25073.1 | - | 1 | 132 | |||
M26710.1 | - | 1 | 179 |
