siRNA Details
Gene Symbol: Crhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247542.2 | XP_006247604.1 | 10 | 2301 | ||
GenBank mRNA | EU012436.1 | - | 11 | 1027 | ||
EU012437.1 | - | 10 | 876 | |||
EU012438.1 | - | 11 | 1095 | |||
L24096.1 | - | 11 | 1108 | |||
L25438.1 | - | 11 | 1030 |
