siRNA Details
Gene Symbol: Adcy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031007.1 | NP_112269.1 | 4 | 882 | ||
GenBank mRNA | M80550.1 | - | 4 | 882 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adcy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031007.1 | NP_112269.1 | 4 | 882 | ||
GenBank mRNA | M80550.1 | - | 4 | 882 |