siRNA Details
Gene Symbol: Cspg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031022.1 | NP_112284.1 | 3 | 650 | ||
GenBank mRNA | X56541.2 | - | 3 | 650 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cspg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031022.1 | NP_112284.1 | 3 | 650 | ||
GenBank mRNA | X56541.2 | - | 3 | 650 |