siRNA Details
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024268.2 | NP_001019439.1 | 16 | 1654 | ||
GenBank mRNA | AF276774.1 | - | 16 | 1684 | ||
BC093604.1 | - | 16 | 1638 | 7317599 |
Gene Symbol: LOC100911727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759094.1 | XP_008757316.1 | 12 | 1323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
