siRNA Details
Gene Symbol: Gnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030987.2 | NP_112249.2 | 7, 8 | 577 | ||
XM_006239517.2 | XP_006239579.1 | 7, 8 | 729 | |||
XM_006239518.2 | XP_006239580.1 | 7, 8 | 729 | |||
XM_006239519.2 | XP_006239581.1 | 7, 8 | 729 | |||
XM_006239520.2 | XP_006239582.1 | 7, 8 | 729 | |||
XM_008764335.1 | XP_008762557.1 | 6, 7 | 612 | |||
GenBank mRNA | AY552805.1 | - | 5, 6 | 429 | ||
BC078809.1 | - | 7, 8 | 577 | 7123318 | ||
DQ120483.1 | - | 2, 3 | 182 | |||
DQ120484.1 | - | 2, 3 | 182 | |||
U34958.1 | - | 5, 6 | 429 | |||
U88324.1 | - | 5, 6 | 466 |
