siRNA Details
Gene Symbol: Crhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247542.2 | XP_006247604.1 | 10, 11 | 2344 | ||
GenBank mRNA | EU012436.1 | - | 11, 12 | 1070 | ||
EU012437.1 | - | 10, 11 | 919 | |||
EU012438.1 | - | 11, 12 | 1138 | |||
L24096.1 | - | 11, 12 | 1151 | |||
L25438.1 | - | 11, 12 | 1073 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
