siRNA Details
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031029.1 | NP_112291.1 | 7 | 772 | ||
XM_008767594.1 | XP_008765816.1 | 8 | 1358 | |||
XM_008767595.1 | XP_008765817.1 | 8 | 1364 | |||
GenBank mRNA | FQ222085.1 | - | 8 | 939 | ||
U95368.1 | - | 7 | 772 |
