siRNA Details
Gene Symbol: Rps6ka1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031107.1 | NP_112369.1 | 2, 3 | 257 | ||
XM_006239084.2 | XP_006239146.1 | 1, 2 | 577 | |||
GenBank mRNA | M99169.1 | - | 2, 3 | 257 |
