siRNA Details
Gene Symbol: Actb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031144.3 | NP_112406.1 | 3 | 425 | ||
GenBank mRNA | AF122902.1 | - | 1 | 146 | ||
BC063166.1 | - | 3 | 428 | 6920838 | ||
EF156276.1 | - | 1 | 147 | |||
FQ231503.1 | - | 3 | 427 | |||
FQ232063.1 | - | 3 | 427 | |||
FQ232650.1 | - | 3 | 427 | |||
KJ696744.1 | - | 3 | 401 |

More Information
Other Species Targeted

Gene Ontology Categories:
