siRNA Details
Gene Symbol: Crisp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011710.2 | NP_001011710.1 | 4, 5 | 286 | ||
XM_006244643.2 | XP_006244705.1 | 3, 4 | 269 | |||
GenBank mRNA | AB009662.1 | - | 3, 4 | 149 | ||
BC078799.1 | - | 4, 5 | 235 | 7112769 | ||
CK469282.1 | - | 4, 5 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
