siRNA Details
Gene Symbol: Mdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031151.2 | NP_112413.2 | 3, 4 | 430 | ||
GenBank mRNA | BC063165.1 | - | 3, 4 | 430 | 6918633 | |
DQ402951.1 | - | 2, 3 | 264 | |||
FQ213494.1 | - | 3, 4 | 383 | |||
FQ215385.1 | - | 3, 4 | 390 | |||
FQ215647.1 | - | 3, 4 | 391 | |||
FQ216782.1 | - | 3, 4 | 421 | |||
FQ217232.1 | - | 3, 4 | 382 | |||
FQ224847.1 | - | 3, 4 | 327 | |||
FQ224979.1 | - | 3, 4 | 409 | |||
X04240.1 | - | 3, 4 | 371 |
