siRNA Details
Gene Symbol: EPHB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004444.4 | NP_004435.3 | 3 | 823 | ||
XM_017011816.1 | XP_016867305.1 | 3 | 426 | |||
GenBank mRNA | AB209644.1 | - | 2 | 1533 | ||
AY056048.1 | - | 3 | 417 | |||
BC052804.1 | - | 3 | 500 | 6584384 | ||
EU826608.1 | - | 3 | 332 | |||
EU826609.1 | - | 3 | 332 | |||
EU826610.1 | - | 3 | 332 | |||
U07695.1 | - | 3 | 417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
