siRNA Details
Gene Symbol: Mdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031151.2 | NP_112413.2 | 9 | 1104 | ||
GenBank mRNA | BC063165.1 | - | 9 | 1104 | 6918633 | |
FQ213494.1 | - | 9 | 1056 | |||
FQ213817.1 | - | 9 | 1019 | |||
FQ215385.1 | - | 9 | 1064 | |||
FQ215647.1 | - | 9 | 1065 | |||
FQ216782.1 | - | 9 | 1095 | |||
FQ217232.1 | - | 9 | 1056 | |||
FQ224847.1 | - | 9 | 1001 | |||
X04240.1 | - | 9 | 1045 |
