siRNA Details
Gene Symbol: Rab11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031152.2 | NP_112414.1 | 3 | 468 | ||
GenBank mRNA | BC085727.1 | - | 3 | 468 | 7189729 | |
FQ214239.1 | - | 3 | 532 | |||
FQ220492.1 | - | 3 | 540 | |||
M75153.1 | - | 3 | 403 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_017382, AK132072, AB232605, AK014268, D50500, and others.
Gene Ontology Categories:
