siRNA Details
Gene Symbol: Crisp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011710.2 | NP_001011710.1 | 9 | 778 | ||
XM_006244643.2 | XP_006244705.1 | 8 | 761 | |||
GenBank mRNA | AB009662.1 | - | 8 | 641 | ||
BC078799.1 | - | 9 | 727 | 7112769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
