siRNA Details
Gene Symbol: Fads2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031344.2 | NP_112634.1 | 3 | 537 | ||
XM_008760244.1 | XP_008758466.1 | 3 | 2046 | |||
GenBank mRNA | AB021980.1 | - | 3 | 467 | ||
BC081776.1 | - | 3 | 537 | 7128630 | ||
FQ210893.1 | - | 3 | 466 | |||
HQ909027.1 | - | 3 | 371 |
