siRNA Details
Gene Symbol: Tsc22d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031345.1 | NP_112635.1 | 2, 3 | 380 | ||
XM_006234249.2 | XP_006234311.1 | 3, 4 | 647 | |||
XM_006234250.2 | XP_006234312.1 | 3, 4 | 561 | |||
XM_006234251.2 | XP_006234313.1 | 3, 4 | 596 | |||
XM_008773446.1 | XP_008771668.1 | 2, 3 | 229 | |||
XM_008773447.1 | XP_008771669.1 | 3, 4 | 500 | |||
GenBank mRNA | AB025431.1 | - | 2, 3 | 380 | ||
BC061979.1 | - | 2, 3 | 378 | 5598145 |
