siRNA Details
Gene Symbol: CCNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005190.3 | NP_005181.2 | 1 | 302 | ||
XM_006715594.1 | XP_006715657.1 | 1 | 461 | |||
XM_011536232.2 | XP_011534534.1 | 1 | 461 | |||
XM_017011436.1 | XP_016866925.1 | 1 | 461 | |||
GenBank mRNA | AB125137.1 | - | 1 | 89 | ||
AK222768.1 | - | 1 | 116 | |||
AK223247.1 | - | 1 | 103 | |||
AK291914.1 | - | 1 | 111 | |||
AK298558.1 | - | 1 | 148 | |||
BC010135.2 | - | 1 | 135 | 4106819 | ||
BC026272.1 | - | 1 | 116 | |||
BC041123.1 | - | 1 | 285 | 5759974 | ||
BC056153.1 | - | 1 | 124 | 6157106 | ||
M74091.1 | - | 1 | 103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
