siRNA Details
Gene Symbol: St3gal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031337.2 | NP_112627.2 | 5 | 898 | ||
XM_006236676.1 | XP_006236738.1 | 4 | 559 | |||
XM_006236677.1 | XP_006236739.1 | 5 | 701 | |||
XM_006236679.2 | XP_006236741.1 | 4 | 692 | |||
XM_006236680.1 | XP_006236742.1 | 4 | 559 | |||
XM_006236682.1 | XP_006236744.1 | 3 | 386 | |||
XM_006236684.2 | XP_006236746.1 | 3 | 580 | |||
XM_008762996.1 | XP_008761218.1 | 4 | 721 | |||
XM_008762997.1 | XP_008761219.1 | 3 | 609 | |||
GenBank mRNA | AB018049.1 | - | 4 | 564 | ||
BC078798.1 | - | 5 | 898 | 7106217 |
