siRNA Details
Gene Symbol: EPHB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280794.2 | NP_001267723.2 | 16 | 3242 | ||
NM_001280795.2 | NP_001267724.2 | 13 | 2614 | |||
NM_004445.5 | NP_004436.4 | 14 | 2683 | |||
NR_104001.2 | 16 | 2494 | ||||
XM_006715881.2 | XP_006715944.1 | 17 | 3678 | |||
XM_011515879.1 | XP_011514181.1 | 18 | 3717 | |||
XM_011515880.1 | XP_011514182.1 | 17 | 3695 | |||
XM_011515881.2 | XP_011514183.1 | 17 | 3676 | |||
XM_011515882.2 | XP_011514184.1 | 17 | 3588 | |||
GenBank mRNA | AL832533.1 | - | 4 | 1545 | ||
BC056159.1 | - | 16 | 3111 | |||
BC110606.1 | - | 13 | 2608 | 40028676 | ||
BC110607.1 | - | 14 | 2677 | 40028687 | ||
D83492.1 | - | 17 | 3254 | |||
EU054308.1 | - | 12 | 2620 |
