siRNA Details
Gene Symbol: Neu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031522.3 | NP_113710.2 | 3 | 618 | ||
GenBank mRNA | AB035722.1 | - | 3 | 476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
