siRNA Details
Gene Symbol: Bcl2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033670.1 | NP_001028842.1 | 2 | 561 | ||
NM_001033672.1 | NP_001028844.1 | 2 | 718 | |||
NM_031535.2 | NP_113723.2 | 2 | 546 | |||
XM_006235264.2 | XP_006235326.1 | 2 | 560 | |||
XM_006235265.2 | XP_006235327.1 | 2 | 1373 | |||
GenBank mRNA | AY141038.1 | - | 2 | 718 | ||
BC094213.1 | - | 2 | 561 | 7314572 | ||
S76513.1 | - | 1 | 402 | |||
U34963.1 | - | 1 | 402 | |||
U72349.1 | - | 2 | 546 | |||
U72350.1 | - | 1 | 454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
