siRNA Details
Gene Symbol: Sv2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031593.1 | NP_113781.1 | 2 | 381 | ||
GenBank mRNA | AF060174.1 | - | 2 | 381 |
Gene Symbol: RGD1559896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109134.1 | NP_001102604.1 | 3 | 917 | ||
XM_008772642.1 | XP_008770864.1 | 3 | 934 | |||
XM_008772643.1 | XP_008770865.1 | 3 | 817 | |||
GenBank mRNA | FQ212098.1 | - | 3 | 659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
