siRNA Details
Gene Symbol: IL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185156.1 | NP_001172085.1 | 3 | 436 | ||
NM_001185157.1 | NP_001172086.1 | 3 | 436 | |||
NM_006850.3 | NP_006841.1 | 3 | 433 | |||
XM_011509101.2 | XP_011507403.1 | 2 | 328 | |||
XM_017000121.1 | XP_016855610.1 | 2 | 326 | |||
GenBank mRNA | AY641441.1 | - | 3 | 435 | ||
BC009681.1 | - | 3 | 409 | 3878467 | ||
BP222545.1 | - | 3 | 433 | |||
BT007156.1 | - | 2 | 161 | |||
U16261.1 | - | 3 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
