siRNA Details
Gene Symbol: Prg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031619.2 | NP_113807.2 | 3 | 241 | ||
GenBank mRNA | CB703061.1 | - | 2 | 236 | ||
D50568.1 | - | 3 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
