siRNA Details
Gene Symbol: Nr4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031628.1 | NP_113816.1 | 6, 7 | 2167 | ||
XM_008763702.1 | XP_008761924.1 | 7, 8 | 3961 | |||
XM_008763703.1 | XP_008761925.1 | 6, 7 | 3889 | |||
GenBank mRNA | D38530.1 | - | 6, 7 | 2167 | ||
DQ268830.1 | - | 6, 7 | 2167 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_015743, AF191211, BC128307, BC128308, BC068150, and others.
Gene Ontology Categories:
