siRNA Details
Gene Symbol: Kcnab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031652.1 | NP_113840.1 | 1 | 618 | ||
XM_008767862.1 | XP_008766084.1 | 1 | 656 | |||
GenBank mRNA | X76723.1 | - | 1 | 618 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010599, AK083134, AK049316, AK046194, BC017518, and others.
Gene Ontology Categories:
