siRNA Details
Gene Symbol: ERBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005862.2 | NP_001005862.1 | 6 | 804 | ||
NM_001289936.1 | NP_001276865.1 | 7 | 855 | |||
NM_001289937.1 | NP_001276866.1 | 3 | 579 | |||
NM_001289938.1 | NP_001276867.1 | 6 | 804 | |||
NM_004448.3 | NP_004439.2 | 3 | 579 | |||
NR_110535.1 | 3 | 642 | ||||
GenBank mRNA | AF177761.2 | - | 3 | 330 | ||
AK131568.1 | - | 1 | 1010 | |||
AK295195.1 | - | 7 | 855 | |||
AK296908.1 | - | 3 | 394 | |||
AK313683.1 | - | 3 | 533 | |||
BC080193.1 | - | 3 | 540 | |||
BC110392.1 | - | 6 | 769 | |||
BC167147.1 | - | 3 | 321 | 100013444 | ||
BI755753.1 | - | 6 | 769 | |||
KJ534831.1 | - | 3 | 318 | |||
KJ534832.1 | - | 3 | 318 | |||
KJ534964.1 | - | 3 | 318 | |||
M11730.1 | - | 3 | 468 | |||
X03363.1 | - | 3 | 492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
