siRNA Details
Gene Symbol: Pla2g2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031598.3 | NP_113786.3 | 3, 4 | 239 | ||
XM_006239146.2 | XP_006239208.1 | 4, 5 | 405 | |||
GenBank mRNA | AF365363.1 | - | 2, 3 | 183 | ||
AY651027.1 | - | 2, 3 | 183 | |||
D00523.1 | - | 3, 4 | 223 | |||
FQ227448.1 | - | 3, 4 | 239 | |||
FQ234749.1 | - | 4, 5 | 337 | |||
M25148.1 | - | 3, 4 | 239 | |||
X74364.1 | - | 1, 2 | 119 |
