siRNA Details
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031606.1 | NP_113794.1 | 2, 3 | 172 | ||
GenBank mRNA | AF017185.1 | - | 2, 3 | 172 | ||
AF455569.1 | - | 2, 3 | 172 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008960, U92437, AK088717, BC021445, AK076980, and others.
Gene Ontology Categories:
