siRNA Details
Gene Symbol: Nr4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031628.1 | NP_113816.1 | 8 | 2381 | ||
XM_008763702.1 | XP_008761924.1 | 9 | 4175 | |||
XM_008763703.1 | XP_008761925.1 | 8 | 4103 | |||
GenBank mRNA | D38530.1 | - | 8 | 2381 | ||
DQ268830.1 | - | 8 | 2381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
