siRNA Details
Gene Symbol: ERBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005862.2 | NP_001005862.1 | 7 | 1004 | ||
NM_001289936.1 | NP_001276865.1 | 8 | 1055 | |||
NM_001289937.1 | NP_001276866.1 | 4 | 779 | |||
NM_001289938.1 | NP_001276867.1 | 7 | 1004 | |||
NM_004448.3 | NP_004439.2 | 4 | 779 | |||
NR_110535.1 | 4 | 842 | ||||
GenBank mRNA | AF177761.2 | - | 4 | 530 | ||
AK131568.1 | - | 2 | 1210 | |||
AK295195.1 | - | 8 | 1055 | |||
AK296908.1 | - | 4 | 594 | |||
AK313683.1 | - | 4 | 733 | |||
BC080193.1 | - | 4 | 740 | |||
BC110392.1 | - | 7 | 969 | |||
BC167147.1 | - | 4 | 521 | 100013444 | ||
KJ534831.1 | - | 4 | 518 | |||
KJ534964.1 | - | 4 | 518 | |||
M11730.1 | - | 4 | 668 | |||
X03363.1 | - | 4 | 692 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
