siRNA Details
Gene Symbol: Txnrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF108213.1 | - | 5 | 459 | ||
AF220760.1 | - | 5 | 513 | |||
AF220761.1 | - | 5 | 491 | |||
U63923.2 | - | 5 | 508 |

More Information
Other Species Targeted

