siRNA Details
Gene Symbol: Nr1h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031627.2 | NP_113815.2 | 3, 4 | 514 | ||
XM_006234533.2 | XP_006234595.1 | 4, 5 | 692 | |||
XM_006234534.1 | XP_006234596.1 | 4, 5 | 633 | |||
XM_006234535.2 | XP_006234597.1 | 4, 5 | 834 | |||
GenBank mRNA | BC088750.1 | - | 3, 4 | 514 | 7314121 | |
U11685.1 | - | 3, 4 | 521 |
