siRNA Details
Gene Symbol: Scn7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031686.1 | NP_113874.1 | 25 | 5006 | ||
XM_006234296.2 | XP_006234358.1 | 25 | 5217 | |||
XM_006234298.2 | XP_006234360.1 | 24 | 5109 | |||
GenBank mRNA | M96578.1 | - | 2 | 1195 | ||
Y09164.1 | - | 25 | 5006 |
