siRNA Details
Gene Symbol: Rps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031706.1 | NP_113894.1 | 4 | 261 | ||
GenBank mRNA | BC082802.1 | - | 3 | 245 | 7302521 | |
BC166878.1 | - | 4 | 261 | 6920981 | ||
FQ221315.1 | - | 4 | 261 | |||
FQ222655.1 | - | 4 | 298 | |||
FQ222951.1 | - | 4 | 259 | |||
FQ223011.1 | - | 4 | 261 | |||
FQ223286.1 | - | 4 | 263 | |||
FQ229267.1 | - | 4 | 261 | |||
X06423.1 | - | 4 | 261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
