siRNA Details
Gene Symbol: Ddr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031764.3 | NP_113952.2 | 15 | 2398 | ||
XM_006250226.2 | XP_006250288.1 | 16 | 5695 | |||
XM_006250227.2 | XP_006250289.1 | 18 | 3030 | |||
XM_006250228.2 | XP_006250290.1 | 17 | 2811 | |||
XM_006250229.2 | XP_006250291.1 | 17 | 2759 | |||
XM_008769761.1 | XP_008767983.1 | 17 | 2773 | |||
XM_008769762.1 | XP_008767984.1 | 17 | 2705 | |||
XM_008769763.1 | XP_008767985.1 | 17 | 3011 | |||
GenBank mRNA | AF016247.1 | - | 2 | 367 | ||
BC161961.1 | - | 15 | 2398 | 9027893 |

More Information
Other Species Targeted

Gene Ontology Categories:
