siRNA Details
Gene Symbol: Slc1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032065.1 | NP_114454.1 | 3, 4 | 495 | ||
XM_008765182.1 | XP_008763404.1 | 3, 4 | 669 | |||
GenBank mRNA | AY286330.1 | - | 3, 4 | 489 | ||
BC098912.1 | - | 3, 4 | 519 | 7315936 | ||
U80915.1 | - | 1, 2 | 125 | |||
U89608.1 | - | 3, 4 | 495 |
