siRNA Details
Gene Symbol: Ncoa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031822.1 | NP_114010.1 | 2 | 131 | ||
XM_006237744.2 | XP_006237806.1 | 4 | 629 | |||
XM_006237745.2 | XP_006237807.1 | 4 | 232 | |||
XM_006237747.2 | XP_006237809.1 | 3 | 1321 | |||
XM_008763516.1 | XP_008761738.1 | 3 | 1514 | |||
GenBank mRNA | AF136943.1 | - | 2 | 131 |
