siRNA Details
Gene Symbol: Nsfl1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031981.2 | NP_114187.1 | 3 | 371 | ||
XM_006235254.2 | XP_006235316.1 | 3 | 338 | |||
GenBank mRNA | AB002086.1 | - | 3 | 371 | ||
BC072464.1 | - | 3 | 268 | 7096463 | ||
FQ228953.1 | - | 3 | 303 | |||
Y10769.1 | - | 3 | 268 |

More Information
Other Species Targeted

Gene Ontology Categories:
